siRNA Details
Gene Symbol: Jak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190830.1 | NP_001177759.1 | 1 | 186 | ||
NM_010589.6 | NP_034719.2 | 1 | 186 | |||
GenBank mRNA | AK088365.1 | - | 1 | 38 | ||
AK142178.1 | - | 1 | 24 | |||
AK164520.1 | - | 1 | 26 | |||
BC027234.1 | - | 17 | ||||
L40172.1 | - | 1 | 47 |
Gene Symbol: Gm9933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087256.1 | - | 1 | 108 | ||
AK162065.1 | - | 1 | 109 |
