siRNA Details
Gene Symbol: Jak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190830.1 | NP_001177759.1 | 1 | 157 | ||
NM_010589.6 | NP_034719.2 | 1 | 157 |
Gene Symbol: Gm9933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087256.1 | - | 1 | 137 | ||
AK162065.1 | - | 1 | 138 |
