siRNA Details
Gene Symbol: Tmem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033759.2 | NP_001028931.1 | 20 | 3932 | ||
NM_031997.4 | NP_114386.3 | 21 | 4014 | |||
GenBank mRNA | AK129352.1 | - | 15 | 2233 | ||
BC019745.1 | - | 12 | 1655 | 3675225 | ||
BC076570.1 | - | 20 | 3538 | 6830791 | ||
BC089353.1 | - | 20 | 3914 | 6410437 | ||
BI666286.1 | - | 4 | 550 |
