siRNA Details
Gene Symbol: Setdb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163641.1 | NP_001157113.1 | 21 | 3929 | ||
NM_001163642.1 | NP_001157114.1 | 21 | 3888 | |||
NM_018877.3 | NP_061365.3 | 21 | 3891 | |||
GenBank mRNA | AF091628.1 | - | 21 | 3842 | ||
AK088590.1 | - | 14 | 2880 | |||
AK122198.1 | - | 17 | 4394 | |||
AK132202.1 | - | 21 | 3864 | |||
BC007176.1 | - | 8 | 3021 | |||
BC060173.1 | - | 21 | 3883 | |||
BC079537.1 | - | 20 | 4921 | 6410215 |
