siRNA Details
Gene Symbol: Aff4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033565.2 | NP_291043.1 | 20 | 3660 | ||
GenBank mRNA | AF190449.1 | - | 20 | 3602 | ||
AK033163.1 | - | 10 | 1480 | |||
AK053034.1 | - | 8 | 690 | |||
BC028557.1 | - | 2 | 426 | |||
BC138999.1 | - | 20 | 3333 | 8862021 | ||
BC145490.1 | - | 19 | 3246 | 9054053 |

More Information
Other Species Targeted

Gene Ontology Categories:
