siRNA Details
Gene Symbol: Rnf111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033604.2 | NP_291082.1 | 15 | 3299 | ||
GenBank mRNA | AF330197.1 | - | 15 | 3295 | ||
AK036351.1 | - | 14 | 3010 | |||
AK048110.1 | - | 14 | 3041 | |||
AK137148.1 | - | 13 | 2613 | |||
AK164502.1 | - | 6 | 696 | |||
AK169999.1 | - | 14 | 3381 | |||
BC011463.1 | - | 9 | 1707 | |||
BC026485.1 | - | 6 | 636 | |||
BC054842.1 | - | 5 | 538 | |||
BC069835.1 | - | 15 | 3131 | 30094289 |
