siRNA Details
Gene Symbol: Dcaf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153555.2 | NP_705783.1 | 6, 7 | 1177 | ||
GenBank mRNA | AK035701.1 | - | 6, 7 | 1076 | ||
AK146825.1 | - | 6, 7 | 1104 | |||
AK162622.1 | - | 6, 7 | 1199 | |||
AK164799.1 | - | 6, 7 | 1104 | |||
AK167059.1 | - | 6, 7 | 1053 | |||
BC023704.1 | - | 7, 8 | 1118 | 5345701 | ||
BC023804.1 | - | 7, 8 | 1231 | 5352941 |
