siRNA Details
Gene Symbol: Hemgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133294.1 | NP_579828.1 | 3 | 390 | ||
XM_008763693.1 | XP_008761915.1 | 3 | 466 | |||
XM_008763694.1 | XP_008761916.1 | 4 | 534 | |||
GenBank mRNA | AJ302650.2 | - | 3 | 390 | ||
BC078739.1 | - | 4 | 490 | 7111835 |
