siRNA Details
Gene Symbol: C5ar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053619.1 | NP_446071.1 | 2 | 912 | ||
GenBank mRNA | AB003042.1 | - | 2 | 912 | ||
AF090996.1 | - | 1 | 627 | |||
BC078770.1 | - | 2 | 895 | 7121456 | ||
Y09613.1 | - | 2 | 896 |
