siRNA Details
Gene Symbol: Tmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053671.2 | NP_446123.2 | 2 | 375 | ||
XM_008763143.1 | XP_008761365.1 | 2 | 1897 | |||
GenBank mRNA | AF107843.1 | - | 2 | 298 | ||
BF415257.1 | - | 2 | 375 | |||
CK470592.1 | - | 1 | 51 |
