siRNA Details
Gene Symbol: Aurkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053749.1 | NP_446201.1 | 2 | 198 | ||
XM_006246569.2 | XP_006246631.1 | 2 | 216 | |||
XM_008767757.1 | XP_008765979.1 | 2 | 113 | |||
XM_008767758.1 | XP_008765980.1 | 2 | 59 | |||
GenBank mRNA | BC097297.1 | - | 2 | 116 | 7454285 | |
D89731.1 | - | 2 | 198 |
