siRNA Details
Gene Symbol: Ccnc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100472.1 | NP_001093942.1 | 1, 2 | 132 | ||
XM_006237937.2 | XP_006237999.1 | 1, 2 | 216 | |||
GenBank mRNA | BC161908.1 | - | 1, 2 | 202 | ||
D14013.1 | - | 1, 2 | 132 |
