siRNA Details
Gene Symbol: Mapk8ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053777.1 | NP_446229.1 | 7 | 1796 | ||
GenBank mRNA | AF092450.1 | - | 6 | 1531 | ||
AF108959.1 | - | 6 | 1734 | |||
AF109772.1 | - | 6 | 1643 | |||
AF109773.1 | - | 6 | 1706 | |||
AF109774.1 | - | 7 | 1796 | |||
DQ377223.1 | - | 6 | 1606 |
