siRNA Details
Gene Symbol: Wnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002823.2 | NP_001002823.2 | 7 | 2095 | ||
NM_001199095.1 | NP_001186024.1 | 7 | 2095 | |||
NM_053794.2 | NP_446246.2 | 7 | 2095 | |||
XM_008763201.1 | XP_008761423.1 | 7 | 2161 | |||
XM_008763202.1 | XP_008761424.1 | 7 | 2161 | |||
XM_008763203.1 | XP_008761425.1 | 7 | 2161 | |||
XM_008763204.1 | XP_008761426.1 | 7 | 2161 | |||
XM_008763205.1 | XP_008761427.1 | 7 | 2161 | |||
XM_008763206.1 | XP_008761428.1 | 7 | 2161 | |||
XM_008763207.1 | XP_008761429.1 | 7 | 2161 | |||
XM_008763208.1 | XP_008761430.1 | 7 | 2161 | |||
XM_008763209.1 | XP_008761431.1 | 7 | 2161 | |||
XM_008763210.1 | XP_008761432.1 | 7 | 2161 | |||
XM_008763211.1 | XP_008761433.1 | 7 | 2161 | |||
XM_008763212.1 | XP_008761434.1 | 7 | 2161 | |||
XM_008763213.1 | XP_008761435.1 | 7 | 2161 | |||
XM_008763214.1 | XP_008761436.1 | 4 | 1315 | |||
XM_008763215.1 | XP_008761437.1 | 7 | 2161 | |||
GenBank mRNA | AF227741.1 | - | 7 | 2095 |
