siRNA Details
Gene Symbol: Kidins220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053795.1 | NP_446247.1 | 30 | 4235 | ||
XM_006239923.2 | XP_006239985.1 | 31 | 4739 | |||
XM_006239924.2 | XP_006239986.1 | 31 | 4736 | |||
XM_006239925.2 | XP_006239987.1 | 32 | 4429 | |||
XM_006239926.2 | XP_006239988.1 | 31 | 4724 | |||
XM_006239927.2 | XP_006239989.1 | 32 | 4426 | |||
XM_006239928.2 | XP_006239990.1 | 32 | 4411 | |||
XM_006239929.2 | XP_006239991.1 | 30 | 4706 | |||
XM_006239931.2 | XP_006239993.1 | 30 | 4682 | |||
XM_006239932.2 | XP_006239994.1 | 30 | 4667 | |||
XM_006239933.2 | XP_006239995.1 | 29 | 4648 | |||
XM_006239934.2 | XP_006239996.1 | 29 | 4633 | |||
XM_006239935.2 | XP_006239997.1 | 28 | 4600 | |||
XM_006239936.2 | XP_006239998.1 | 27 | 4486 | |||
XM_006239937.2 | XP_006239999.1 | 28 | 4177 | |||
GenBank mRNA | AF239045.2 | - | 30 | 4235 | ||
AF313464.1 | - | 27 | 3980 |
