siRNA Details
Gene Symbol: Crnkl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053797.2 | NP_446249.1 | 1 | 733 | ||
GenBank mRNA | AF245018.1 | - | 1 | 723 | ||
BC085718.1 | - | 1 | 733 | 7190212 |
Gene Symbol: LOC102548514 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235165.2 | XP_006235227.1 | 6 | 780 |
