siRNA Details
Gene Symbol: Nrxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053817.2 | NP_446269.2 | 13 | 3442 | ||
XM_008764731.1 | XP_008762953.1 | 11 | 2683 | |||
GenBank mRNA | L14851.1 | - | 13 | 3442 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nrxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053817.2 | NP_446269.2 | 13 | 3442 | ||
XM_008764731.1 | XP_008762953.1 | 11 | 2683 | |||
GenBank mRNA | L14851.1 | - | 13 | 3442 |