siRNA Details
Gene Symbol: Lct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053841.1 | NP_446293.1 | 8 | 2691 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053841.1 | NP_446293.1 | 8 | 2691 |