siRNA Details
Gene Symbol: Mapk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053842.1 | NP_446294.1 | 7 | 1102 | ||
XM_006248658.2 | XP_006248720.1 | 7 | 1139 | |||
XM_006248659.2 | XP_006248721.1 | 7 | 1139 | |||
XM_008768848.1 | XP_008767070.1 | 7 | 1139 | |||
GenBank mRNA | M64300.1 | - | 7 | 1102 |
