siRNA Details
Gene Symbol: Scg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053856.2 | NP_446308.1 | 2 | 415 | ||
GenBank mRNA | U02983.1 | - | 2 | 417 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Scg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053856.2 | NP_446308.1 | 2 | 415 | ||
GenBank mRNA | U02983.1 | - | 2 | 417 |