siRNA Details
Gene Symbol: Dync1i2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270624.1 | NP_001257553.1 | 16 | 2095 | ||
NM_001270625.1 | NP_001257554.1 | 15 | 2035 | |||
NM_053880.2 | NP_446332.2 | 17 | 2113 | |||
XM_006234344.2 | XP_006234406.1 | 16 | 1872 | |||
XM_006234346.2 | XP_006234408.1 | 15 | 1811 | |||
GenBank mRNA | BC078764.1 | - | 16 | 1841 | 7113840 | |
U39044.1 | - | 17 | 1829 | |||
U39045.1 | - | 16 | 1811 | |||
U39046.1 | - | 15 | 1751 |
