siRNA Details
Gene Symbol: Pgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229825.2 | XP_006229887.1 | 4 | 895 | ||
XM_006229830.2 | XP_006229892.1 | 4 | 896 | |||
XM_006229831.2 | XP_006229893.1 | 4 | 882 | |||
XM_006229832.2 | XP_006229894.1 | 4 | 882 | |||
XM_006229834.2 | XP_006229896.1 | 4 | 882 | |||
XM_006229837.2 | XP_006229899.1 | 4 | 872 | |||
XM_006229838.2 | XP_006229900.1 | 4 | 872 | |||
GenBank mRNA | AB236144.1 | - | 3 | 347 | ||
U57715.1 | - | 4 | 864 |
