siRNA Details
Gene Symbol: Cgrrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053899.1 | NP_446351.1 | 4, 5 | 676 | ||
GenBank mRNA | BC078763.1 | - | 3, 4 | 822 | 7107668 | |
FQ218613.1 | - | 4, 5 | 691 | |||
U66471.1 | - | 4, 5 | 676 |
