siRNA Details
Gene Symbol: Kynu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053902.2 | NP_446354.1 | 12 | 1074 | ||
XM_006234138.2 | XP_006234200.1 | 11 | 990 | |||
GenBank mRNA | BC078762.1 | - | 12 | 1057 | 7128911 | |
U68168.1 | - | 12 | 1075 |
