siRNA Details
Gene Symbol: Kynu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053902.2 | NP_446354.1 | 9 | 810 | ||
XM_006234138.2 | XP_006234200.1 | 8 | 726 | |||
XM_008761695.1 | XP_008759917.1 | 9 | 873 | |||
GenBank mRNA | BC078762.1 | - | 9 | 793 | 7128911 | |
U68168.1 | - | 9 | 811 |
