siRNA Details
Gene Symbol: Inpp4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053917.1 | NP_446369.1 | 25 | 3054 | ||
XM_008772462.1 | XP_008770684.1 | 25 | 2897 | |||
XM_008772465.1 | XP_008770687.1 | 24 | 2846 | |||
GenBank mRNA | U96921.1 | - | 25 | 3054 |
