siRNA Details
Gene Symbol: Slc7a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053929.1 | NP_446381.1 | 13 | 1483 | ||
XM_006228875.2 | XP_006228937.1 | 14 | 1585 | |||
XM_006228876.2 | XP_006228938.1 | 14 | 1965 | |||
XM_006228877.2 | XP_006228939.1 | 13 | 1844 | |||
XM_006228878.2 | XP_006228940.1 | 10 | 1150 | |||
XM_008759103.1 | XP_008757325.1 | 13 | 1869 | |||
XM_008759104.1 | XP_008757326.1 | 13 | 1524 | |||
GenBank mRNA | AB029559.1 | - | 13 | 1483 | ||
BC081750.1 | - | 12 | 1899 | |||
BC098909.1 | - | 13 | 1473 | 7109606 |
