siRNA Details
Gene Symbol: Ctrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054009.1 | NP_446461.1 | 7 | 680 | ||
GenBank mRNA | AB020757.1 | - | 7 | 680 | ||
FQ234836.1 | - | 7 | 694 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ctrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054009.1 | NP_446461.1 | 7 | 680 | ||
GenBank mRNA | AB020757.1 | - | 7 | 680 | ||
FQ234836.1 | - | 7 | 694 |