siRNA Details
Gene Symbol: Prpsap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057131.1 | NP_476472.1 | 8, 9 | 823 | ||
XM_006246434.2 | XP_006246496.1 | 8, 9 | 824 | |||
XM_006246435.2 | XP_006246497.1 | 9, 10 | 1030 | |||
XM_006246436.2 | XP_006246498.1 | 8, 9 | 815 | |||
XM_006246437.2 | XP_006246499.1 | 7, 8 | 777 | |||
XM_006246438.2 | XP_006246500.1 | 7, 8 | 698 | |||
XM_006246439.2 | XP_006246501.1 | 6, 7 | 785 | |||
XM_006246440.2 | XP_006246502.1 | 8, 9 | 898 | |||
XM_008767764.1 | XP_008765986.1 | 7, 8 | 843 | |||
XM_008767765.1 | XP_008765987.1 | 6, 7 | 725 | |||
GenBank mRNA | BC078755.1 | - | 5, 6 | 558 | 7113134 | |
D84434.1 | - | 8, 9 | 823 | |||
FQ212452.1 | - | 8, 9 | 771 |
