siRNA Details
Gene Symbol: Cflar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033864.2 | NP_001029036.1 | 3, 4 | 560 | ||
NM_057138.2 | NP_476479.1 | 3, 4 | 560 | |||
XM_006244946.2 | XP_006245008.1 | 3, 4 | 1127 | |||
XM_006244947.2 | XP_006245009.1 | 3, 4 | 1234 | |||
XM_006244948.2 | XP_006245010.1 | 2, 3 | 2748 | |||
XM_006244949.2 | XP_006245011.1 | 2, 3 | 563 | |||
XM_008767071.1 | XP_008765293.1 | 3, 4 | 653 | |||
GenBank mRNA | AF244366.1 | - | 1, 2 | 350 | ||
BC089781.1 | - | 560 | 7376876 |
