siRNA Details
Gene Symbol: Slc4a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270860.1 | NP_001257789.1 | 7 | 971 | ||
NM_001270861.1 | NP_001257790.1 | 7 | 1010 | |||
NM_058211.2 | NP_478118.1 | 7 | 1010 | |||
GenBank mRNA | AF069511.1 | - | 7 | 971 | ||
AF070475.1 | - | 7 | 1010 | |||
AF080106.1 | - | 7 | 1010 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: JX310364, AF218295, HM624051.
Gene Ontology Categories:
