siRNA Details
Gene Symbol: Ryk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080402.1 | NP_536327.1 | 11 | 1189 | ||
XM_008766473.1 | XP_008764695.1 | 11 | 1180 | |||
GenBank mRNA | AB073721.1 | - | 4 | 266 | ||
AY669340.1 | - | 11 | 1189 | |||
BC099232.1 | - | 4 | 247 |
