siRNA Details
Gene Symbol: Faf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130406.1 | NP_569090.1 | 13 | 1432 | ||
GenBank mRNA | AF293459.1 | - | 13 | 1432 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007983, BC065098, U39643, AK053752.
Gene Ontology Categories:
