siRNA Details
Gene Symbol: Arhgef5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_342676.7 | XP_342677.4 | 2 | 1485 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Arhgef5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_342676.7 | XP_342677.4 | 2 | 1485 |