siRNA Details
Gene Symbol: Selenbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080892.1 | NP_543168.1 | 5 | 616 | ||
GenBank mRNA | AB036799.1 | - | 5 | 616 | ||
BC074008.1 | - | 5 | 626 | 7100467 | ||
FQ219337.1 | - | 5 | 596 |
Gene Symbol: LOC103689947 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761279.1 | XP_008759501.1 | 616 |
