siRNA Details
Gene Symbol: Slc36a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130415.1 | NP_569099.1 | 11 | 1512 | ||
XM_008767657.1 | XP_008765879.1 | 10 | 3611 | |||
XM_008767659.1 | XP_008765881.1 | 12 | 2194 | |||
GenBank mRNA | AF361239.1 | - | 11 | 1512 |
