siRNA Details
Gene Symbol: Tmprss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130424.2 | NP_569108.2 | 3 | 167 | ||
XM_008768547.1 | XP_008766769.1 | 3 | 283 | |||
XM_008768548.1 | XP_008766770.1 | 3 | 214 | |||
XM_008768549.1 | XP_008766771.1 | 3 | 396 | |||
GenBank mRNA | AB073550.1 | - | 1 | 65 | ||
BC061712.1 | - | 3 | 167 | 5598010 |
