siRNA Details
Gene Symbol: Acot12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130747.1 | NP_570103.1 | 12 | 1255 | ||
XM_008760628.1 | XP_008758850.1 | 13 | 1750 | |||
GenBank mRNA | AB040609.1 | - | 12 | 1255 |
