siRNA Details
Gene Symbol: Ppp3cc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134367.1 | NP_599194.1 | 8 | 975 | ||
XM_008770813.1 | XP_008769035.1 | 8 | 1158 | |||
XM_008770814.1 | XP_008769036.1 | 8 | 1157 | |||
XM_008770815.1 | XP_008769037.1 | 8 | 1592 | |||
XM_008770816.1 | XP_008769038.1 | 9 | 969 | |||
XM_008770817.1 | XP_008769039.1 | 8 | 1506 | |||
GenBank mRNA | BC079027.1 | - | 8 | 975 | 7114037 | |
GQ376198.1 | - | 8 | 884 | |||
GQ376199.1 | - | 8 | 884 | |||
M58442.1 | - | 3 | 191 |
