siRNA Details
Gene Symbol: A2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012488.2 | NP_036620.2 | 11 | 1192 | ||
GenBank mRNA | AY921651.1 | - | 11 | 1151 | ||
BC098922.1 | - | 11 | 1192 | 7458629 | ||
FQ223246.1 | - | 11 | 1214 | |||
FQ228339.1 | - | 11 | 1391 | |||
J02635.1 | - | 11 | 1214 |
Gene Symbol: LOC100911545 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749828.2 | XP_003749876.1 | 1498 |
