siRNA Details
Gene Symbol: Acly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111095.1 | NP_001104565.1 | 25, 26 | 3142 | ||
NM_016987.2 | NP_058683.2 | 26, 27 | 3172 | |||
XM_006247254.2 | XP_006247316.1 | 26, 27 | 3165 | |||
GenBank mRNA | BC100618.1 | - | 25, 26 | 3134 | 7934668 | |
J05210.1 | - | 26, 27 | 3120 |
