siRNA Details
Gene Symbol: Acly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111095.1 | NP_001104565.1 | 21, 22 | 2584 | ||
NM_016987.2 | NP_058683.2 | 22, 23 | 2614 | |||
XM_006247254.2 | XP_006247316.1 | 22, 23 | 2607 | |||
GenBank mRNA | BC100618.1 | - | 21, 22 | 2576 | 7934668 | |
J05210.1 | - | 22, 23 | 2562 |
