siRNA Details
Gene Symbol: Aldoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012497.1 | NP_036629.1 | 7 | 824 | ||
GenBank mRNA | BC099749.1 | - | 7 | 810 | 5599032 | |
FQ212739.1 | - | 1 | 91 | |||
FQ213260.1 | - | 1 | 91 | |||
FQ214420.1 | - | 7 | 850 | |||
FQ228893.1 | - | 7 | 824 | |||
X06984.1 | - | 7 | 824 |
