siRNA Details
Gene Symbol: Cacna1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012517.2 | NP_036649.2 | 33, 34 | 4425 | ||
XM_006237159.2 | XP_006237221.1 | 35, 36 | 4384 | |||
XM_006237170.2 | XP_006237232.1 | 33, 34 | 4270 | |||
XM_006237171.2 | XP_006237233.1 | 33, 34 | 4270 | |||
XM_006237172.2 | XP_006237234.1 | 33, 34 | 4270 | |||
XM_006237175.2 | XP_006237237.1 | 34, 35 | 4778 | |||
XM_008763216.1 | XP_008761438.1 | 34, 35 | 6287 | |||
GenBank mRNA | AF394938.1 | - | 33, 34 | 4276 | ||
AF394939.1 | - | 33, 34 | 4276 | |||
AF394940.1 | - | 33, 34 | 4276 | |||
AY974797.2 | - | 35, 36 | 4542 | |||
DQ538522.1 | - | 35, 36 | 4244 | |||
M59786.1 | - | 33, 34 | 4425 | |||
M67515.1 | - | 33, 34 | 4419 | |||
M67516.1 | - | 33, 34 | 4904 | |||
U31815.1 | - | 6, 7 | 602 |
