siRNA Details
Gene Symbol: Chgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012526.2 | NP_036658.2 | 4 | 1596 | ||
XM_006235072.2 | XP_006235134.1 | 5 | 1506 | |||
GenBank mRNA | AF019974.1 | - | 4 | 1396 | ||
BC128743.1 | - | 4 | 1415 | 8370781 |
