siRNA Details
Gene Symbol: Grin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270602.1 | NP_001257531.1 | 10 | 1634 | ||
NM_001270603.1 | NP_001257532.1 | 10 | 1634 | |||
NM_001270605.1 | NP_001257534.1 | 9 | 1571 | |||
NM_001270606.1 | NP_001257535.1 | 10 | 1634 | |||
NM_001270608.1 | NP_001257537.1 | 10 | 1634 | |||
NM_001270610.1 | NP_001257539.1 | 9 | 1571 | |||
NM_001287423.1 | NP_001274352.1 | 9 | 1571 | |||
NM_017010.2 | NP_058706.1 | 9 | 1571 | |||
XM_006233555.2 | XP_006233617.1 | 10 | 1640 | |||
XM_006233556.2 | XP_006233618.1 | 9 | 1577 | |||
GenBank mRNA | U08261.1 | - | 9 | 1482 | ||
U08262.1 | - | 9 | 1482 | |||
U08263.1 | - | 10 | 1521 | |||
U08264.1 | - | 10 | 1521 | |||
U08265.1 | - | 9 | 1482 | |||
U08266.1 | - | 10 | 1521 | |||
U08267.1 | - | 9 | 1482 | |||
U08268.1 | - | 10 | 1521 | |||
U11418.1 | - | 9 | 1387 | |||
X63255.1 | - | 9 | 1512 | |||
X65227.1 | - | 10 | 1310 |
