siRNA Details
Gene Symbol: Grin2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012573.3 | NP_036705.3 | 13 | 3014 | ||
GenBank mRNA | D13211.1 | - | 12 | 2807 | ||
M91561.1 | - | 13 | 3014 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Grin2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012573.3 | NP_036705.3 | 13 | 3014 | ||
GenBank mRNA | D13211.1 | - | 12 | 2807 | ||
M91561.1 | - | 13 | 3014 |