siRNA Details
Gene Symbol: Hp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012582.2 | NP_036714.2 | 5 | 699 | ||
GenBank mRNA | AF476963.1 | - | 5 | 730 | ||
AY325231.1 | - | 6 | 827 | |||
BC089816.1 | - | 5 | 699 | 7321960 | ||
FQ209575.1 | - | 3 | 604 | |||
FQ209592.1 | - | 5 | 751 | |||
FQ209684.1 | - | 5 | 751 | |||
FQ211144.1 | - | 5 | 753 | |||
FQ219122.1 | - | 5 | 751 | |||
FQ219413.1 | - | 2 | 509 | |||
FQ219443.1 | - | 5 | 751 | |||
FQ219547.1 | - | 5 | 722 | |||
FQ219598.1 | - | 5 | 721 | |||
K01933.1 | - | 2 | 463 |
