siRNA Details
Gene Symbol: Slco2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022667.2 | NP_073158.2 | 4, 5 | 706 | ||
XM_006243655.1 | XP_006243717.1 | 5, 6 | 688 | |||
GenBank mRNA | M64862.1 | - | 4, 5 | 707 |
