siRNA Details
Gene Symbol: Phrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139093.1 | NP_620793.1 | 16 | 4805 | ||
XM_006230510.2 | XP_006230572.1 | 17 | 5018 | |||
XM_006230511.2 | XP_006230573.1 | 17 | 4976 | |||
XM_006230513.2 | XP_006230575.1 | 17 | 5015 | |||
XM_006230514.2 | XP_006230576.1 | 17 | 4973 | |||
XM_006230515.2 | XP_006230577.1 | 17 | 4943 | |||
XM_006230516.2 | XP_006230578.1 | 17 | 4898 | |||
XM_006230517.2 | XP_006230579.1 | 17 | 4895 | |||
GenBank mRNA | U49057.1 | - | 16 | 4805 |
